missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat PTMA Protein

A DNA sequence encoding the Rattus norvegicus (Rat) PTMA, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9354-ye-500ug

 View more versions of this product

Product Code. 16068171

  • 25885.00 SEK / 500µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_068508.1
PTMA Protein
Research Use Only
Ptma
Recombinant Protein
Yeast
SDAAVDTSSE ITTKDLKEKK EVVEEAENGR DAPANGNAQN EENGEQEADN EVDEEEEEGG EEEEEEEEGD GEEEDGDEDE EAEAPTGKRV AEDDEDDDVE TKKQKKTDED D
Greater than 90% as determined by SDS-PAGE.
100359583
500 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant Rat PTMA Protein > 500μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.