missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant S. aureus Adapter protein MecA Protein Product Code.: 16027860

enQuireBio™ Recombinant S. aureus Adapter protein MecA Protein

Product Code. 16027860
10 μg, 10µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16027860

Brand: enQuireBio™ QP7252ec10ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Staphylococcus aureus (strain MW2) Adapter protein MecA, was expressed in the hosts and tags indicated.

Specifications

Accession Number WP_001217728.1
Gene ID (Entrez) 31213749
Name S. aureus Adapter protein MecA Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol mecA
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity S. aureus
Species E. coli
Protein Tag His-SUMO
Sequence MRIERVDDTT VKLFITYSDI EARGFSREDL WTNRKRGEEF FWSMMDEINE EEDFVVEGPL WIQVHAFEKG VEVTISKSKN EDMMNMSDDD ATDQFDEQVQ ELLAQTLEGE DQLEELFEQR TKEKEAQGSK RQKSSARKNT RTIIVKFNDL EDVINYAYHS NPITTEFEDL LYMVDGTYYY AVHFDSHVDQ EVINDSYSQL LEFAYPTDRT EVYLNDYAKI IMSHNVTAQV RRYFPETTE
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant S. aureus Adapter protein MecA Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.