missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus Clumping factor A Protein

Product Code. 16010631
1 mg, 1mg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy
This item is not returnable. View return policy

A DNA sequence encoding the Staphylococcus aureus Clumping factor A, was expressed in the hosts and tags indicated.

Specifications

Accession Number WP_001056174.1
Name S. aureus Clumping factor A Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol clfA
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity S. aureus
Species E. coli
Protein Tag His-SUMO
Sequence GKDITNQLTN VTVGIDSGDT VYPHQAGYVK LNYGFSVPNS AVKGDTFKIT VPKELNLNGV TSTAKVPPIM AGDQVLANGV IDSDGNVIYT FTDYVDTKEN VTANITMPAY IDPENVTKTG NVTLTTGIGS TTANKTVLVD YEKYGKFYNL SIKGTIDQID KTNNTYRQTI YVNPSGDNVI APVLTGNLKP NTDSNALIDQ QNTSIKVYKV DNAADLSESY FVNPENFEDV TNSVNITFPN PNQYKVEFNT PDDQITTPYI VVVNGHIDPN SKGDLALRST LYGYDSRFVW RSMSWDNEVA FNNGSGSGDG IDKPVVPEQP DEPGEIEPIP E
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant S. aureus Clumping factor A Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.