Learn More
enQuireBio™ Recombinant S. aureus Staphylococcal secretory antigen ssaA2 Protein
A DNA sequence encoding the Staphylococcus aureus (strain Mu50 / ATCC 700699) Staphylococcal secretory antigen ssaA2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8015-ec-50ug
This item is not returnable.
View return policy
Specifications
WP_000717381.1 | |
50 ÎĽg | |
Not determined. | |
Full Length | |
S. aureus | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
S. aureus Staphylococcal secretory antigen ssaA2 Protein | |
Research Use Only | |
ssaA2 | |
Recombinant Protein | |
E. coli | |
SEQDNYGYNP NDPTSYSYTY TIDAQGNYHY TWKGNWHPSQ LNQDNGYYSY YYYNGYNNYN NYNNGYSYNN YSRYNNYSNN NQSYNYNNYN SYNTNSYRTG GLGASYSTSS NNVQVTTTMA PSSNGRSISS GYTSGRNLYT SGQCTYYVFD RVGGKIGSTW GNASNWANAA ARAGYTVNNT PKAGAIMQTT QGAYGHVAYV ESVNSNGSVR VSEMNYGYGP GVVTSRTISA SQAAGYNFIH | |
Greater than 90% as determined by SDS-PAGE. |
Product Content Correction
Product Title
enQuireBio™ Recombinant S. aureus Staphylococcal secretory antigen ssaA2 Protein > 50μg
Spot an opportunity for improvement?Share a Content Correction