Learn More
enQuireBio™ Recombinant S. aureus Staphylococcal secretory antigen ssaA2 Protein
A DNA sequence encoding the Staphylococcus aureus (strain Mu50 / ATCC 700699) Staphylococcal secretory antigen ssaA2, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP8015-ec-10ug
 View more versions of this product
Product Code. 16092161
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
WP_000717381.1
10 ÎĽg
Not determined.
Full Length
S. aureus
His-SUMO
Tris-based buffer, 50% glycerol
S. aureus Staphylococcal secretory antigen ssaA2 Protein
Research Use Only
ssaA2
Recombinant Protein
E. coli
SEQDNYGYNP NDPTSYSYTY TIDAQGNYHY TWKGNWHPSQ LNQDNGYYSY YYYNGYNNYN NYNNGYSYNN YSRYNNYSNN NQSYNYNNYN SYNTNSYRTG GLGASYSTSS NNVQVTTTMA PSSNGRSISS GYTSGRNLYT SGQCTYYVFD RVGGKIGSTW GNASNWANAA ARAGYTVNNT PKAGAIMQTT QGAYGHVAYV ESVNSNGSVR VSEMNYGYGP GVVTSRTISA SQAAGYNFIH
Greater than 90% as determined by SDS-PAGE.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
enQuireBio™ Recombinant S. aureus Staphylococcal secretory antigen ssaA2 Protein > 10ÎĽg