Learn More
enQuireBio™ Recombinant S. cerevisiae Cystathionine beta-synthase Protein
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
Specifications
Specifications
Accession Number | NP_011671.3 |
Gene ID (Entrez) | 853059 |
Name | S. cerevisiae Cystathionine beta-synthase Protein |
Quantity | 500 μg |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | CYS4 |
Protein Length | Full Length |
Product Type | Recombinant Protein |
Cross Reactivity | S. cerevisiae |
Show More |
Product Content Correction
Product Title
enQuireBio™ Recombinant S. cerevisiae Cystathionine beta-synthase Protein >
Spot an opportunity for improvement?Share a Content Correction