Learn More
enQuireBio™ Recombinant S. cerevisiae Cystathionine beta-synthase Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Cystathionine beta-synthase, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP8802-ye-50ug
 View more versions of this product
Product Code. 16076551
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
NP_011671.3
S. cerevisiae Cystathionine beta-synthase Protein
Research Use Only
CYS4
Recombinant Protein
Yeast
MTKSEQQADS RHNVIDLVGN TPLIALKKLP KALGIKPQIY AKLELYNPGG SIKDRIAKSM VEEAEASGRI HPSRSTLIEP TSGNTGIGLA LIGAIKGYRT IITLPEKMSN EKVSVLKALG AEIIRTPTAA AWDSPESHIG VAKKLEKEIP GAVILDQYNN MMNPEAHYFG TGREIQRQLE DLNLFDNLRA VVAGAGTGGT ISGISKYLKE QNDKIQIVGA DPFGSILAQP ENLNKTDITD YKVEGIGYDF VPQVLDRKLI DVWYKTDDKP SFKYARQLIS NEGVLVGGSS GSAFTAVVKY CEDHPELTED DVIVAIFPDS IRSYLTKFVD DEWLKKNNLW DDDVLARFDS SKLEASTTKY ADVFGNATVK DLHLKPVVSV KETAKVTDVI KILKDNGFDQ LPVLTEDGKL SGLVTLSELL RKLSINNSNN DNTIKGKYLD FKKLNNFNDV SSYNENKSGK KKFIKFDENS KLSDLNRFFE KNSSAVITDG LKPIHIVTKM DLLSYLA
Greater than 90% as determined by SDS-PAGE.
853059
50 ÎĽg
Not determined.
Full Length
S. cerevisiae
His
Tris-based buffer, 50% glycerol
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
enQuireBio™ Recombinant S. cerevisiae Cystathionine beta-synthase Protein > 50ÎĽg