missing translation for 'onlineSavingsMsg'
Learn More
enQuireBio™ Recombinant S. cerevisiae Cystathionine beta-synthase Protein Product Code.: 16056551

enQuireBio™ Recombinant S. cerevisiae Cystathionine beta-synthase Protein

Product Code. 16056551
200 μg, 200µg
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16056551

Brand: enQuireBio™ QP8802ye200ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Cystathionine beta-synthase, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_011671.3
Gene ID (Entrez) 853059
Name S. cerevisiae Cystathionine beta-synthase Protein
Quantity 200 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol CYS4
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity S. cerevisiae
Species Yeast
Protein Tag His
Sequence MTKSEQQADS RHNVIDLVGN TPLIALKKLP KALGIKPQIY AKLELYNPGG SIKDRIAKSM VEEAEASGRI HPSRSTLIEP TSGNTGIGLA LIGAIKGYRT IITLPEKMSN EKVSVLKALG AEIIRTPTAA AWDSPESHIG VAKKLEKEIP GAVILDQYNN MMNPEAHYFG TGREIQRQLE DLNLFDNLRA VVAGAGTGGT ISGISKYLKE QNDKIQIVGA DPFGSILAQP ENLNKTDITD YKVEGIGYDF VPQVLDRKLI DVWYKTDDKP SFKYARQLIS NEGVLVGGSS GSAFTAVVKY CEDHPELTED DVIVAIFPDS IRSYLTKFVD DEWLKKNNLW DDDVLARFDS SKLEASTTKY ADVFGNATVK DLHLKPVVSV KETAKVTDVI KILKDNGFDQ LPVLTEDGKL SGLVTLSELL RKLSINNSNN DNTIKGKYLD FKKLNNFNDV SSYNENKSGK KKFIKFDENS KLSDLNRFFE KNSSAVITDG LKPIHIVTKM DLLSYLA
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
enQuireBio™ Recombinant S. cerevisiae Cystathionine beta-synthase Protein >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.