missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant S. cerevisiae GTP-binding protein YPT1 Protein
A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) GTP-binding protein YPT1, was expressed in the hosts and tags indicated.
3070.00 SEK - 38570.00 SEK
Specifications
Accession Number | NP_116615.1 |
---|---|
Gene ID (Entrez) | 850505 |
Name | S. cerevisiae GTP-binding protein YPT1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16008320
|
enQuireBio™
QP7341-EC-100UG |
100 μg |
11125.00 SEK
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16028320
|
enQuireBio™
QP7341-EC-1MG |
1 mg |
38570.00 SEK
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16018320
|
enQuireBio™
QP7341-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16038320
|
enQuireBio™
QP7341-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16048320
|
enQuireBio™
QP7341-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16058320
|
enQuireBio™
QP7341-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Spezifikation
NP_116615.1 | |
S. cerevisiae GTP-binding protein YPT1 Protein | |
Not determined. | |
Full Length | |
S. cerevisiae | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
850505 | |
Research Use Only | |
YPT1 | |
Recombinant Protein | |
E. coli | |
MNSEYDYLFK LLLIGNSGVG KSCLLLRFSD DTYTNDYIST IGVDFKIKTV ELDGKTVKLQ IWDTAGQERF RTITSSYYRG SHGIIIVYDV TDQESFNGVK MWLQEIDRYA TSTVLKLLVG NKCDLKDKRV VEYDVAKEFA DANKMPFLET SALDSTNVED AFLTMARQIK ESMSQQNLNE TTQKKEDKGN VNLKGQSLTN TGGGCC | |
Greater than 90% as determined by SDS-PAGE. |