missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Sheep IL6 / Interleukin-6 Protein
A DNA sequence encoding the Ovis aries (Sheep) IL6 / Interleukin-6 Protein, was expressed in the hosts and tags indicated.
3490.00 SEK - 31480.00 SEK
Specifications
Accession Number | NP_001009392.1 |
---|---|
Gene ID (Entrez) | 443406 |
Name | IL6 / Interleukin-6 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16021730
|
enQuireBio™
QP6225-ye-10ug |
10 ÎĽg | N/A | ||||||
16061730
|
enQuireBio™
QP6225-ye-50ug |
50 ÎĽg | N/A | ||||||
16011730
|
enQuireBio™
QP6225-ye-100ug |
100 ÎĽg |
12935.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16041730
|
enQuireBio™
QP6225-ye-200ug |
200 ÎĽg | N/A | ||||||
16051730
|
enQuireBio™
QP6225-ye-500ug |
500 ÎĽg |
31245.00 SEK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16031730
|
enQuireBio™
QP6225-ye-1mg |
1 mg | N/A | ||||||
Specifications
NP_001009392.1 | |
IL6 / Interleukin-6 Protein | |
Not determined. | |
Full Length | |
Sheep | |
His | |
Tris-based buffer, 50% glycerol |
443406 | |
Research Use Only | |
IL6 | |
Recombinant Protein | |
Yeast | |
GPLGEDFKND TTPSRLLLTT PEKTEALIKH IVDKISAIRK EICEKNDECE NSKETLAENK LKLPKMEEKD GCFQSGFNQA ICLIKTTAGL LEYQIYLDFL QNEFEGNQET VMELQSSIRT LIQILKEKIA GLITTPATHT DMLEKMQSSN EWVKNAKVII ILRSLENFLQ FSLRAIRMK | |
Greater than 80% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Sheep IL6 / Interleukin-6 Protein