Learn More
enQuireBio™ Recombinant Zebrafish Metallothionein-2 Protein
A DNA sequence encoding the Danio rerio (Zebrafish) (Brachydanio rerio) Metallothionein-2, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP9826-ye-500ug
 View more versions of this product
Product Code. 16068791
  • 31245.00 SEK / 500µg
This item is not returnable. View return policy
Specifications
NP_001124525.2
Metallothionein-2 Protein
Research Use Only
mt2
Recombinant Protein
Yeast
MDPCECAKTG TCNCGATCKC TNCQCTTCKK SCCSCCPSGC SKCASGCVCK GNSCGSSCCQ
Greater than 90% as determined by SDS-PAGE.
100174951
500 μg
Not determined.
Full Length
Zebrafish
His
Tris-based buffer, 50% glycerol
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Zebrafish Metallothionein-2 Protein > 500μg