Learn More
Thermo Scientific™ Remel™ OADC Enrichment
Remel OADC Enrichment is used in Middlebrook Media.
Brand:  Thermo Scientific™ R450603
1004.58 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 13436039
  • 1075.00 SEK / 100mL
Only null left
Estimated Shipment: 22-09-2025
Description
Remel OADC Enrichment is used in Middlebrook Media.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
OADC Enrichment
Liquid
OADC Enrichment
100 mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number281948 Certificate TypeCertificate of Analysis Date14/07/2025 Catalog Number
Lot Number273963 Certificate TypeCertificate of Analysis Date26/06/2025 Catalog Number
Lot Number258972 Certificate TypeCertificate of Analysis Date14/06/2025 Catalog Number
Lot Number266041 Certificate TypeCertificate of Analysis Date14/06/2025 Catalog Number
Lot Number260338 Certificate TypeCertificate of Analysis Date14/06/2025 Catalog Number
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Remel™ OADC Enrichment > OADC Enrichment; 100mL; 1 Ea.