Learn More
Trajan Scientific and Medical™ Replacement Plungers for Agilent Syringes
Click to view available options
Volume (Metric):
10 μL
100 μL
Product Type:
Fixed Needle
Removable Needle
Unit Size:
Each
Pack of 2
Description
Designed to match the specific syringe barrel to maintain a gas tight seal
Specifications
Specifications
Description | Replacement plunger |
Diameter (Metric) Outer | 0.63/0.47 mm |
Product Type | Removable Needle |
Volume (Metric) | 10 μL |
Needle Point Style | Cone |
For Use With (Equipment) | Gas Tight Syringe |
Product Content Correction
Product Title
Trajan Scientific and Medical™ Replacement Plungers for Agilent Syringes >
Spot an opportunity for improvement?Share a Content Correction