missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REPS2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20699-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The antibody is specific to REPS2.
The product of this gene is part of a protein complex that regulates the endocytosis of growth factor receptors. The encoded protein directly interacts with a GTPase activating protein that functions downstream of the small G protein Ral. Its expression can negatively affect receptor internalization and inhibit growth factor signaling. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
REPS2 | |
Polyclonal | |
Unconjugated | |
REPS2 | |
Partner of RalBP1, POB1, RalBP1 interacting protein 2, REPS2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
194590, 9185 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.56 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q80XA6, Q8NFH8 | |
REPS2 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |