missing translation for 'onlineSavingsMsg'
Learn More
Learn More
REPS2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1355.00 SEK - 3665.00 SEK
Specifications
Antigen | REPS2 |
---|---|
Concentration | 0.56 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16850275
|
Proteintech
20699-1-AP-20UL |
20 μL |
1355.00 SEK
20µL |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16840275
|
Proteintech
20699-1-AP-150UL |
150 μL |
3665.00 SEK
150µL |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The antibody is specific to REPS2.
The product of this gene is part of a protein complex that regulates the endocytosis of growth factor receptors. The encoded protein directly interacts with a GTPase activating protein that functions downstream of the small G protein Ral. Its expression can negatively affect receptor internalization and inhibit growth factor signaling. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
REPS2 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
Q80XA6, Q8NFH8 | |
194590, 9185 | |
Peptide | |
Primary | |
-20°C | |
REPS2 |
0.56 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Partner of RalBP1, POB1, RalBP1 interacting protein 2, REPS2 | |
REPS2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |