missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Retinal S antigen Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23546-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
S-antigen, also identified as arrestin, is a protein involved in the regulation of phototransduction.Specifications
Retinal S antigen | |
Polyclonal | |
Unconjugated | |
SAG | |
48 kDa protein, Retinal S antigen, Rod photoreceptor arrestin, S AG, S arrestin, SAG | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20215, 6295 | |
-20°C | |
Liquid |
Western Blot | |
0.52 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P10523, P20443 | |
SAG | |
Retinal S antigen Fusion Protein Ag20318 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |