Learn More
Brady™ Retro Reflective All-weather Labels
Increase safety and visibility with the B-584 highly-reflective label material. Print labels almost anywhere high visibility is needed with a compact Brady™ handheld label printer.
Brand:  Brady™ 170811
 View more versions of this product
Product Code. 17971930
  • ONLINE EXCLUSIVE PRICE
    585.00 SEK
    551.14 SEK / Paket med 1
    Save 33.86 SEK
    5% Av
I lager
Description
  • Ideal for safety applications such as on personnel, fire suppression equipment, and first aid cabinets or anywhere extra visibility is required
  • Resistant to abrasions, weather, solvents and extreme temperatures
  • Pressure-sensitive acrylic adhesive withstands prolonged exposure to tough conditions
  • Able to withstand outdoor conditions for 4 to 6 years on average
Specifications
Specifications
Not Autoclavable
Permanent Acrylic
White
Continuous
Top
Label
Component and equipment labelling;Facility identification;General identification;Safety identification
-40°C
1 Label(s) / Box
19.05 mm (W) x 6.10 m (L)
-40°C to 70°C
0.14 mm
20 ft.
0.75 in.
19.05 mm
M4C-750-584
2011/65/EU, 2015/863
None
Gloss
Plastic
Thermal Transfer
Non-sterile
BMP41, BMP51, BMP53, M511, M410, M510
M4C-750-584
1
-40°F to 158°F
0.0055 in
19.05 mm (W) x 6.10 m (L)
6.10 m
19.05 mm
0.5 in.