Learn More
RGS14 Mouse anti-Human, Unconjugated, Clone: 1C1B2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67394-1-IG-20UL
Description
The RGS14 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RGS14. This antibody recognizes human, mouse, rat antigen. The RGS14 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
RGS14 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, ELISA | |
1C1B2 | |
Unconjugated | |
20 μL | |
10636 | |
IgG1 |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.