missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RGS17 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12549-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the regulator of G-protein signaling family. This protein contains a conserved, 120 amino acid motif called the RGS domain and a cysteine-rich region. The protein attenuates the signaling activity of G-proteins by binding to activated, GTP-bound G alpha subunits and acting as a GTPase activating protein (GAP), increasing the rate of conversion of the GTP to GDP. This hydrolysis allows the G alpha subunits to bind G beta/gamma subunit heterodimers, forming inactive G-protein heterotrimers, thereby terminating the signal.Specifications
RGS17 | |
Polyclonal | |
Unconjugated | |
RGS17 | |
hRGS17, RGS 17, RGS17, RGSZ2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26575, 56533 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9QZB0, Q9UGC6 | |
RGS17 | |
RGS17 Fusion Protein Ag3399 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RGS17 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction