missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHCG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11949-1-AP-150UL
This item is not returnable.
View return policy
Description
RHCG (Rhesus blood group family type C glycoprotein), also known as RHGK (Rh glycoprotein kidney), PDRC2 (tumor-related protein DRC2) or SLC42A3, is a multi-pass membrane protein with three potential N-glycosylation sites that belongs to the Rh subfamily of the ammonium transporter family. Expressed in a wide variety of tissues with predomit expression in adult and fetal kidney, RHCg localizes to the apical and/or basolateral cell membrane (depending on the species) and is believed to function as a non-erythroid, bidirectional, electroneutral ammonium transporter.Specifications
RHCG | |
Polyclonal | |
Unconjugated | |
RHCG | |
Ammonium transporter Rh type C, C15orf6, CDRC2, PDRC2, Rh family type C glycoprotein, Rh family, C glycoprotein, Rh glycoprotein kidney, Rh type C glycoprotein, RHCG, RHGK, SLC42A3, Tumor related protein DRC2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
293048, 51458, 56315 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q7TNN9, Q9QXP0, Q9UBD6 | |
RHCG | |
RHCG Fusion Protein Ag2493 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RHCG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction