Learn More
RHOC Mouse anti-Human, Unconjugated, Clone: 1G1C1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67542-1-IG-20UL
Description
The RHOC antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RHOC. This antibody recognizes human, mouse, rat antigen. The RHOC antibody has been validated for the following applications: FC, IF, WB, ELISA analysis.
Specifications
RHOC | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunofluorescence, Flow Cytometry, ELISA | |
1G1C1 | |
Unconjugated | |
20 μL | |
389 | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.