missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RHOF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12290-1-AP-150UL
This item is not returnable.
View return policy
Description
Plasma membrane-associated small GTPase which cycles between an active GTP-bound and an inactive GDP-bound state. Causes the formation of thin, actin-rich surface projections called filopodia. Functions cooperatively with CDC42 and Rac to generate additional structures, increasing the diversity of actin-based morphology.Specifications
RHOF | |
Polyclonal | |
Unconjugated | |
RHOF | |
ARHF, Rho family GTPase Rif, Rho in filopodia, RHOF, RIF | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23912, 54509, 690130 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BYP3, Q9HBH0 | |
RHOF | |
RHOF Fusion Protein Ag2943 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RHOF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction