Learn More
RIC8B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17790-1-AP-150UL
 View more versions of this product
Product Code. 16887764
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
Guanine nucleotide exchange factor (GEF), which can activate some, but not all, G-alpha proteins by exchanging bound GDP for free GTP.
Specifications
Specifications
RIC8B
Polyclonal
Unconjugated
RIC8B
hSyn, RIC8, RIC8B
Rabbit
Antigen Affinity Chromatography
RUO
237422, 314681, 55188
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.38 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q80XE1, Q80ZG0, Q9NVN3
Ric8b
RIC8B Fusion Protein Ag11888
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RIC8B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated