Learn More
RIC8B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17790-1-AP-150UL
 View more versions of this product
Product Code. 16887764
  • 4652.90 SEK / 150µL
Description
Guanine nucleotide exchange factor (GEF), which can activate some, but not all, G-alpha proteins by exchanging bound GDP for free GTP.
Specifications
Specifications
RIC8B
Polyclonal
Unconjugated
RIC8B
hSyn, RIC8, RIC8B
Rabbit
Antigen Affinity Chromatography
RUO
237422, 314681, 55188
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.38 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q80XE1, Q80ZG0, Q9NVN3
Ric8b
RIC8B Fusion Protein Ag11888
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
RIC8B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated