missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIMBP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15716-1-AP-150UL
This item is not returnable.
View return policy
Description
The presynaptic active zone mediates synaptic vesicle exocytosis at a synapse. RIM proteins are active zone scaffolding molecules that--among others--mediate vesicle priming and directly or indirectly interact with most other essential presynaptic proteins. RIMBP2 encodes RIMS-binding protein 2 belonging to RIMBP family. RIMBP2 is supposed to be localized at cell membrane especially synaptic plasma membrane. By interacting with RIMS1, RIMS2, CACNA1D and CACNA1B via the SH3 domains, RIMBP2 mediates synaptic transmission as bifunctional linker.Specifications
RIMBP2 | |
Polyclonal | |
Unconjugated | |
RIMBP2 | |
KIAA0318, RBP2, RIM BP2, RIMBP2, RIMS binding protein 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
231760, 23504, 266780 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O15034, Q80U40, Q9JIR1 | |
Rimbp2 | |
RIMBP2 Fusion Protein Ag8390 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RIMBP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction