missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RIPK5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | RIPK5 |
---|---|
Concentration | 0.32 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16879584
|
Proteintech
20102-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16869584
|
Proteintech
20102-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody is specific to DSTYK.
This gene encodes a dual serine/threonine and tyrosine protein kinase which is expressed in multiple tissues. It is thought to function as a regulator of cell death. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
RIPK5 | |
Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
Q6XUX1, Q6XUX2, Q6XUX3 | |
213452, 25778, 304791 | |
RIPK5 Fusion Protein Ag13678 | |
Primary | |
-20°C | |
DSTYK |
0.32 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
DSTYK, Dusty PK, Dusty protein kinase, DustyPK, HDCMD38P, KIAA0472, RIP homologous kinase, RIP5, RIPK5, SgK496, Sugen kinase 496 | |
Dstyk | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RIPK5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech