Learn More
RLBP1L2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21394-1-AP-150UL
 View more versions of this product
Product Code. 16880995
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
Required for normal morphology of late endosomes and/or lysosomes in neurons. Binds phosphatidylinositol 3, 5-bisphosphate (PtdIns(3, 5)P2).
Specifications
Specifications
RLBP1L2
Polyclonal
Unconjugated
CLVS2
C6orf212, C6orf213, Clavesin 2, CLVS2, RLBP1L2
Rabbit
Antigen Affinity Chromatography
RUO
134829, 215890, 361459
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Western Blot
0.28 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
A6JUQ6, Q5SYC1, Q8BG92
Clvs2
RLBP1L2 Fusion Protein Ag16061
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RLBP1L2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated