missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RMND5A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17559-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
RMND5A is a protein coding gene.Specifications
RMND5A | |
Polyclonal | |
Unconjugated | |
RMND5A | |
CTLH, p44CTLH, Protein RMD5 homolog A, RMD5, RMND5A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
64795 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9H871 | |
RMND5A | |
RMND5A Fusion Protein Ag11751 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |