missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNASEH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15606-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
RNASEH1 belongs to the RNase H family. It contains 1 RNase H domain. RNASEH1 is an endonuclease that specifically degrades the RNA of RNA-DNA hybrids.Specifications
RNASEH1 | |
Polyclonal | |
Unconjugated | |
RNASEH1 | |
H1RNA, Ribonuclease H type II, ribonuclease H1, RNase H1, RNASEH1, RNH1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19819, 246243, 298933 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunocytochemistry, Immunoprecipitation | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O60930, O70338, Q5BK46 | |
Rnaseh1 | |
RNASEH1 Fusion Protein Ag7990 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |