missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNF4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17810-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene contains a RING finger motif and acts as a transcription regulator. This protein has been shown to interact with, and inhibit the activity of, TRPS1, a transcription suppressor of GATA-mediated transcription. Transcription repressor ZNF278/PATZ is found to interact with this protein, and thus reduce the enhancement of androgen receptor-dependent transcription mediated by this protein. Studies of the mouse and rat counterparts suggested a role of this protein in spermatogenesis. A pseudogene of this gene is found on chromosome 1.Specifications
RNF4 | |
Polyclonal | |
Unconjugated | |
RNF4 | |
E3 ubiquitin ligase RNF4, Protein SNURF, RES4 26, ring finger protein 4, RNF4, SNURF | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19822, 29274, 6047 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O88846, P78317, Q9QZS2 | |
RNF4 | |
RNF4 Fusion Protein Ag12115 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RNF4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction