missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNFT2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25685-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
TMEM118 is a multi-pass membrane protein, and contains 1 RING-type zinc finger. The function of TMEM118 remains unknown.Specifications
RNFT2 | |
Polyclonal | |
Unconjugated | |
RNFT2 | |
RNFT2, TMEM118 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84900 | |
-20°C | |
Liquid |
Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96EX2 | |
RNFT2 | |
RNFT2 Fusion Protein Ag22313 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |