missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RNFT2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | RNFT2 |
---|---|
Concentration | 0.25 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16824255
|
Proteintech
25685-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16814255
|
Proteintech
25685-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
TMEM118 is a multi-pass membrane protein, and contains 1 RING-type zinc finger. The function of TMEM118 remains unknown.Specifications
RNFT2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
Q96EX2 | |
84900 | |
RNFT2 Fusion Protein Ag22313 | |
Primary | |
-20°C | |
RNFT2 |
0.25 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
RNFT2, TMEM118 | |
RNFT2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |