Learn More
RNH1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10345-1-AP-20UL
 View more versions of this product
Product Code. 16886673
  • 1645.54 SEK / 20µL
Description
RNH1 is the inhibitor of pancreatic RNase and angiogenin. RNH1 may also function in the modulation of cellular activities.
Specifications
Specifications
RNH1
Polyclonal
Unconjugated
RNH1
Placental RNase inhibitor, PRI, RAI, Ribonuclease inhibitor, RNH, RNH1
Rabbit
Antigen Affinity Chromatography
RUO
107702, 6050
-20°C
Liquid
Immunoprecipitation, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P13489, Q91VI7
RNH1
RNH1 Fusion Protein Ag0350
20 μL
Primary
Human, Mouse
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
RNH1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated