Learn More
RNMT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13743-1-AP-20UL
 View more versions of this product
Product Code. 16882114
  • 1645.54 SEK / 20µL
Description
RNMT belongs to the mRNA cap methyltransferase family. RNMT is a mRNA-capping methyltransferase that methylates the N7 position of the added guanosine to the 5′-cap structure of mRNAs. It binds RNA containing 5′-terminal GpppC.
Specifications
Show More
Certificates
Promotions
Promotions
Product Content Correction
Product Title
RNMT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated