Learn More
RNPEP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14764-1-AP-20UL
 View more versions of this product
Product Code. 16823574
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
Exopeptidase which selectively removes arginine and/or lysine residues from the N-terminus of several peptide substrates including Arg(0)-Leu-enkephalin, Arg(0)-Met-enkephalin and Arg(-1)-Lys(0)-somatostatin-14. Can hydrolyze leukotriene A4 (LTA-4) into leukotriene B4 (LTB-4).
Specifications
Specifications
RNPEP
Polyclonal
Unconjugated
RNPEP
Aminopeptidase B, AP B, APB, Arginine aminopeptidase, Arginyl aminopeptidase, RNPEP
Rabbit
Antigen Affinity Chromatography
RUO
215615, 6051, 81761
-20°C
Liquid
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O09175, Q8VCT3, Q9H4A4
RNPEP
RNPEP Fusion Protein Ag6526
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RNPEP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated