Learn More
Bel-Art™ Robotic Sterile Pipette Tips
Designed for use with automated liquid handling equipment
Brand: Bel-Art™ F37860-3100
This item is not returnable.
View return policy
Description
- Sterile, disposable, filtered pipette tips can be used in robotic systems
- Filtered to prevent contamination of pipette arm
- Conductive tip models detect liquid level when pipetting conductive liquids
- Ensures repeatable volume accuracy
- DNase, RNase, pyrogen free, manufactured in Class 8 clean room
- CE marked
Specifications
1000 μL | |
Sterile | |
Not Autoclavable | |
For: Hamilton, Volume: 100 μL, Conductive, Material: carbon filled polypropylene | |
Standard | |
Robotic pipette tips | |
Single-use | |
No |
Filtered | |
Gray | |
CE marked | |
Hanging Tip Rack | |
16 racks x 96 tips | |
Yes | |
Hamilton STAR, STARlet and NIMBUS | |
Carbon-filled Polypropylene |
Product Content Correction
Product Title
Bel-Art™ Robotic Sterile Pipette Tips > For: Hamilton, Volume: 100 μL, Conductive, Material: carbon filled polypropylene
Spot an opportunity for improvement?Share a Content Correction