Learn More
Hamilton™ Robotics Low Bleed High Temperature Septa
Brand:  Hamilton™ 75836
 View more versions of this product
Product Code. 10710844
  • 1880.00 SEK / Pack of 50
Only null left
Estimated Shipment: 17-07-2025
Specifications
12.7 mm
GC
50/Pk.
Low Bleed High Temperature Septa
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Hamilton™ Robotics Low Bleed High Temperature Septa > Outer Diameter: 12.7mm