missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROCK2 Mouse anti-Human, Mouse, Rat, Clone: 1C7B8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66633-1-IG-150UL
Additional Details : Weight : 0.00010kg
Description
This antibody is specific to ROCK2.
The function of this protein remains unknown.Specifications
ROCK2 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O75116, P70336, Q62868 | |
ROCK2 | |
ROCK2 Fusion Protein Ag16330 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG2b |
Immunohistochemistry (Paraffin), Western Blot | |
1C7B8 | |
Unconjugated | |
ROCK2 | |
KIAA0619, p164 ROCK 2, Rho kinase 2, ROCK2 | |
Mouse | |
Protein A | |
RUO | |
19878, 25537, 9475 | |
-20°C | |
Liquid |