missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROCK2 Mouse anti-Human, Mouse, Rat, Clone: 1C7B8, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | ROCK2 |
---|---|
Clone | 1C7B8 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16828695
|
Proteintech
66633-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16818695
|
Proteintech
66633-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody is specific to ROCK2.
The function of this protein remains unknown.Specifications
ROCK2 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
KIAA0619, p164 ROCK 2, Rho kinase 2, ROCK2 | |
ROCK2 | |
IgG2b | |
Protein A | |
Antibody |
1C7B8 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
O75116, P70336, Q62868 | |
19878, 25537, 9475 | |
ROCK2 Fusion Protein Ag16330 | |
Primary | |
-20°C | |
ROCK2 |