Learn More
ROCK2 Mouse anti-Human, Mouse, Rat, Clone: 1C7B8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66633-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16828695
  • 1656.73 SEK / 20µL
Estimated Shipment: 30-05-2024
to see stock.
Description
Description
This antibody is specific to ROCK2.
The function of this protein remains unknown.
Specifications
Specifications
ROCK2
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O75116, P70336, Q62868
ROCK2
ROCK2 Fusion Protein Ag16330
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG2b
Western Blot, Immunohistochemistry (Paraffin)
1C7B8
Unconjugated
ROCK2
KIAA0619, p164 ROCK 2, Rho kinase 2, ROCK2
Mouse
Protein A
RUO
19878, 25537, 9475
-20°C