missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROPN1L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12565-1-AP-150UL
This item is not returnable.
View return policy
Description
Principal Names: ROPN1L; Ropporin-1 like; ASP; AKAP-associated sperm protein; Official Gene Symbol: ROPN1L Gene ID: 83853 (human) Gene Map Locus: 5p15.2 (human) AKAPs act as scaffolding proteinscoordinating phosphatase/kinase signaling at the cytoskeleton. They interact with PKAs and anchor them to specific sub-cellular compartments and participate in multiple cellular processes including growth, metabolism and differentiation. ASP, a 26 kDa protein, is a novel AKAP-interacting protein. It interacts with the amphipathic helix of AKAP3 and facilitates AKAP3-PKA interactions. It consists of a conserved N-terminal AKAP-binding domain and a central cysteine-rich Fz domain. It is specifically expressed in the sperms and might play a significant role in sperm motility and also facilitate cytoskeletal reorganization during spermiogenesis.Specifications
ROPN1L | |
Polyclonal | |
Unconjugated | |
ROPN1L | |
AKAP associated sperm protein, ASP, ROPN1 like protein, ROPN1L, ropporin 1 like, Ropporin 1 like protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
252967, 685646, 83853 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96C74, Q9EQ00 | |
Ropn1l | |
ROPN1L Fusion Protein Ag3178 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ROPN1L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction