Learn More
Hettich™ Rotor Adapter for Carrier no. 1425
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand: Hettich™ 1441
970.30 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Rotor adapter for Hettich Carriers
- For use with Universal 320/320 R Centrifuge
- Universal Centrifuges can be used for both standard and special applications, they are cost-effective and save valuable space in the laboratory
Specifications
Adapter | |
7 x 4 to 10 mL Tubes | |
100 mm | |
7 |
Conical, Blood/Urine | |
16 mm | |
Carrier no 1425 and Swing-out Rotor 1494 for Universal 320/320 R Centrifuge |
Product Content Correction
Product Title
Hettich™ Rotor Adapter for Carrier no. 1425 > 7 x 10mL
Spot an opportunity for improvement?Share a Content Correction