Learn More
Hettich™ Rotor Adapter for Carrier no. 1425
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand:  Hettich™ 1443
624.17 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10347322
  • 687.00 SEK / Each
Description
  • Rotor adapter for Hettich Carriers
  • For use with Universal 320/320 R Centrifuge
  • Universal Centrifuges can be used for both standard and special applications, they are cost-effective and save valuable space in the laboratory
Specifications
Specifications
Adapter
50 mL
115 mm
1
Conical
29 mm
Carrier no 1425 and Swing-out Rotor 1494 for Universal 320/320 R Centrifuge
1
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Hettich™ Rotor Adapter for Carrier no. 1425 > 50mL