Learn More
Hettich™ Rotor Adapter for Carrier no. 1752
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand:  Hettich™ 1783-A
1168.90 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 12200209
  • 1285.00 SEK / Each
Description
  • For use with ROTINA 380/380 R
  • The ROTINA 380's versatility and size make an invaluable addition to any lab. Ideal for clinical, research, food and beverage and industrial applications.
  • Robust and steady high-capacity centrifuge
  • Accessories with bio-containment protect users from aerosols while ensuring optimal safety, for instance, while centrifuging infectious samples
  • Temperature control system with pre-cool function from -20 to +40°C
  • Versa-Rotor™ for multiple applications
  • All-steel construction
  • Quiet Operation
  • Easy-Lift™/Safety-Lock™ Lid
Specifications
Specifications
Rotor Adapter
16 x 2.6 to 7 mL
100 mm
Blood/Urine Tubes
13 mm
ROTINA 380/380 R Centrifuge with Carrier no. 1752
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Hettich™ Rotor Adapter for Carrier no. 1752 > 16 x 7mL