Learn More
Hettich™ Rotor Adapter for Carriers no. 1481, 1490 and 1495
Designed for use with Hettich Rotors. Hettich rotors are optimized for continuous duty and long service life
Brand: Hettich™ 1343
629.84 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Rotor adapter for Hettich carriers
- For use with Rotofix and Universal centrifuges
- Universal Centrifuges can be used for both standard and special applications, they are cost-effective and save valuable space in the laboratory
- ROTOFIX Centrifuges are ideally equipped for tasks in clinical chemistry, it can also play a vital role in cytology
Specifications
Rotor Adapter | |
9 x 3 or 4 mL Tubes | |
88 mm | |
9 |
Round Bottom | |
10 mm | |
Hettich Rotors with Carriers no. 1481, 1490 and 1495 |
Product Content Correction
Product Title
Hettich™ Rotor Adapter for Carriers no. 1481, 1490 and 1495 > 9 x 3 or 4mL
Spot an opportunity for improvement?Share a Content Correction