Learn More
Eppendorf™ Rotor Key for A-4-81
Designed for use with Eppendorf Rotors and Centrifuges
Brand:  Eppendorf™ 5810718309
230.00 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10366652
  • 242.97 SEK / Each
Description
  • Centrifuge rotor key for Eppendorf A-4-81 Series, 4 x 50mL Rotor
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rotor Key
A-4-81 Series; 4 x 50mL Rotor
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Eppendorf™ Rotor Key for A-4-81 > A-4-81 Series; 4 x 50mL Rotor