missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPH3AL Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | RPH3AL |
---|---|
Concentration | 0.16 mg/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16894414
|
Proteintech
15297-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16884414
|
Proteintech
15297-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene plays a direct regulatory role in calcium-ion-dependent exocytosis in both endocrine and exocrine cells and plays a key role in insulin secretion by pancreatic cells. This gene is likely a tumor suppressor. Alternative splicing results in multiple transcript variants encoding distinct isoforms.Specifications
RPH3AL | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human | |
Q9UNE2 | |
9501 | |
RPH3AL Fusion Protein Ag7552 | |
Primary | |
-20°C | |
RPH3AL |
0.16 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
No C2 domains protein, NOC2, Rab effector Noc2, Rabphilin 3A like protein, RPH3AL | |
RPH3AL | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RPH3AL Rabbit anti-Human, Polyclonal, Proteintech