missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL18A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15751-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a protein belonging to the pantothenate kinase family. Pantothenate kinase is a key regulatory enzyme in the biosynthesis of coenzyme A (CoA) in bacteria and mammalian cells. It catalyzes the first committed step in the universal biosynthetic pathway leading to CoA and is itself subject to regulation through feedback inhibition by CoA. This family member is most abundant in muscle but is expressed in all tissues.Specifications
RPL18A | |
Polyclonal | |
Unconjugated | |
RPL18A | |
60S ribosomal protein L18a, ribosomal protein L18a, RPL18A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
290641, 6142, 76808 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P62717, P62718, Q02543 | |
Rpl18a | |
RPL18A Fusion Protein Ag8251 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RPL18A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction