missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL27A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16002-1-AP-150UL
This item is not returnable.
View return policy
Description
Rpl27a gene ontology annotations related to this gene include translation.Specifications
RPL27A | |
Polyclonal | |
Unconjugated | |
RPL27A | |
60S ribosomal protein L27a, ribosomal protein L27a, RPL27A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26451, 293418, 6157 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P14115, P18445, P46776 | |
RPL27A | |
RPL27A Fusion Protein Ag8596 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RPL27A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction