missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL36 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15145-1-AP-150UL
This item is not returnable.
View return policy
Description
The exosome is involved in a multitude of cellular RNA processing and degradation events. DIS3, also known as exosome complex exonuclease RRP44, is a ribonuclease that acts directly in the processing, turnover, and surveillance of a large number of distinct RNA species. DIS3 localizes to both the cytoplasm and the nucleus and contains one PINc domain. It is widely expressed with highest expression in testis and is required for processing of 7S pre-RNA into a mature nuclear complex and, ultimately, for proper mitotic progression. Abnormal expression levels of DIS3 may be associated with colon cancer, suggesting a role for DIS3 in tumorigenesis.Specifications
RPL36 | |
Polyclonal | |
Unconjugated | |
RPL36 | |
60S ribosomal protein L36, ribosomal protein L36, RPL36 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25873, 54217, 58927 | |
-20°C | |
Liquid |
Western Blot | |
0.99 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P39032, P47964, Q9Y3U8 | |
Rpl36 | |
RPL36 Fusion Protein Ag7437 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RPL36 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction